Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00512

Sigma-Aldrich

Phomalone

≥95% (LC/MS-UV)

Synonym(s):

1-[2,4-dihydroxy-3-(2-hydroxyethyl)-6-methoxyphenyl]butan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O5
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=C(C(O)=CC(OC)=C1C(CCC)=O)CCO

InChI

1S/C13H18O5/c1-3-4-9(15)12-11(18-2)7-10(16)8(5-6-14)13(12)17/h7,14,16-17H,3-6H2,1-2H3

InChI key

PTBQWWHUOMDVFS-UHFFFAOYSA-N

General description

Natural product derived from fungal source.

Application

Antifungal metabolite

Biochem/physiol Actions

Shows antagonistic activity in agar culture and on aspen chips against the aspen decay fungus Phellinus tremulae and the blue strain fungus Ophiostoma crassivaginata.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service