Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00316

Sigma-Aldrich

Astragaloside II

≥92% (HPLC)

Synonym(s):

(3β,6α,16β,20R,24S)-3-[(2-O-acetyl-β-D-xylopyranosyl)oxy]-20,24-epoxy-16,25-dihydroxy-9,19-cyclolanostan-6-yl β-D-glucopyranoside, Astrasieversianin VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H70O15
CAS Number:
Molecular Weight:
827.01
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Astragalus membranaceus)

Assay

≥92% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)[C@@H](O[C@]2([H])OC[C@@H](O)[C@H](O)[C@H]2OC(C)=O)CC[C@@]3(C4)[C@@]1([H])[C@@H](O[C@]5([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C[C@]6([H])[C@@]34CC[C@]7(C)[C@]([C@]8(C)O[C@H](C(O)(C)C)CC8)([H])[C@@H](O)C[C@@]67C

InChI

1S/C43H70O15/c1-20(45)54-32-28(48)22(47)18-53-36(32)57-26-10-12-43-19-42(43)14-13-39(6)33(41(8)11-9-27(58-41)38(4,5)52)21(46)16-40(39,7)25(42)15-23(34(43)37(26,2)3)55-35-31(51)30(50)29(49)24(17-44)56-35/h21-36,44,46-52H,9-19H2,1-8H3/t21-,22+,23-,24+,25-,26-,27-,28-,29+,30-,31+,32+,33-,34-,35+,36-,39+,40-,41+,42-,43+/m0/s1

InChI key

AYWNHWGQTMCQIV-PENCHUSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Astragaloside II is a saponin found in the root of Astragalus propinquus, which is known as huáng qí in Traditional Chinese Medicine.

Biochem/physiol Actions

Astragaloside II stimulates osteoblast differentiation resulting in bone formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service