Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702907

Sigma-Aldrich

Anti-SUOX Antibody, clone 6H0C9, Rabbit Monoclonal

Synonym(s):

SUOX, sulfite oxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
clone:
6H0C9, monoclonal
application:
WB
species reactivity:
rat
technique(s):
western blot: 1:500 - 1:2000
citations:

biological source

rabbit

Quality Level

material

colorless

clone

6H0C9, monoclonal

form

liquid

mol wt

60 kDa

species reactivity

rat

concentration

0.80 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

IWVTLGSEVFDVTEFVDLHPGGPSKLMLAAGGPLEPFWALYAVHNQSHVRELLAQYKIGELNPEDKVAPTVETSDPYADDPVRHPALKVNSQRPFNAEPPP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUOX(6821)

General description

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SUOX (P51687).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service