Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5702817

Sigma-Aldrich

Anti-Rev-Erbα/NR1D1 Antibody, clone 9I2H1, Rabbit Monoclonal

Synonym(s):

EAR1, hRev, THRA1, THRAL, ear-1, REVERBA, REVERBalpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

9I2H1, monoclonal

form

liquid

mol wt

75 kDa

species reactivity

rat, human, rat

concentration

1.044mg/mL

technique(s)

western blot: 1:500 - 1:1000

color

colorless

isotype

IgG

immunogen sequence

SPEPTVEDVISQVARAHREIFTYAHDKLGSSPGNFNANHASGSPPATTPHRWENQGCPPAPNDNNTLAAQRHNEALNGLRQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene encodes a transcription factor that is a member of the nuclear receptor subfamily 1. The encoded protein is a ligand-sensitive transcription factor that negatively regulates the expression of core clock proteins. In particular this protein represses the circadian clock transcription factor aryl hydrocarbon receptor nuclear translocator-like protein 1 (ARNTL). This protein may also be involved in regulating genes that function in metabolic, inflammatory and cardiovascular processes.

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 280-360 of human Rev-Erbα/NR1D1 (NP_068370.1).

Application

WB

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service