Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702596

Sigma-Aldrich

Anti-TWEAKR/Fn14/CD266 Antibody, clone 1G4T3, Rabbit Monoclonal

Synonym(s):

CD266, FN14, TWEAKR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

1G4T3, monoclonal

form

liquid

mol wt

17 kDa

species reactivity

rat, human, rat

concentration

1.00 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MDCASCRARPHSDFCLGCAAAPPAPFRLLWPILGGALSLTFVLGLLSGFLVWRRCRRREKFTTPIEETGGEGCPAVALIQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

TNFRSF12A (TNF Receptor Superfamily Member 12A) is a Protein Coding gene. Diseases associated with TNFRSF12A include Multiple Sclerosis and Stroke, Ischemic. Among its related pathways are ERK Signaling and CDK-mediated phosphorylation and removal of Cdc6

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 50-129 of human TWEAKR/Fn14/CD266 (Q9NP84).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service