Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5300516

Sigma-Aldrich

Monoclonal Anti-AXL antibody produced in mouse

clone 7E10, ascites fluid

Synonym(s):

JTK11, UFO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

7E10, monoclonal

mol wt

97.4 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AXL(558)

Immunogen

Purified recombinant extracellular fragment of human AXL fused with hIgGFc tag expressed in HEK293 cell line.
Mouse monoclonal antibody raised against AXL

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy M D'Alfonso et al.
Journal of clinical pathology, 67(8), 690-696 (2014-06-07)
Triple-negative breast cancer comprises a clinically aggressive group of invasive carcinomas. We examined a published gene expression screen of a panel of breast cancer cell lines to identify a potential triple-negative breast cancer-specific gene signature, and attempted to verify our
Dan Li et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 16(4), 511-518 (2014-01-16)
The tyrosine kinase receptor Axl is overexpressed in various types of cancer and correlated with cancer malignancy. Selective Axl blockade reduces tumor growth and metastasis. The purpose of this study was to examine whether the humanized anti-Axl antibody humanized 173
Min-Sik Kim et al.
Molecular & cellular proteomics : MCP, 13(11), 2803-2811 (2014-06-05)
Many patients with pancreatic cancer have metastases to distant organs at the time of initial presentation. Recent studies examining the evolution of pancreatic cancer at the genetic level have shown that clonal complexity of metastatic pancreatic cancer is already initiated
Takehiko Shibata et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3559-3565 (2014-08-27)
Aspergillus fumigatus is a sporulating fungus found ubiquitously in the environment, which is quickly contained in the immunocompetent host but can cause lethal invasive aspergillosis in the immunocompromised host. We have recently demonstrated that Axl (one member of the Tyro3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service