Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5300367

Sigma-Aldrich

Monoclonal Anti-SRC antibody produced in mouse

clone 1F11, ascites fluid

Synonym(s):

ASV, SRC1, c-SRC, p60-Src

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (d)
FACS
IHC
WB
clone:
1F11, monoclonal
species reactivity:
human
citations:
9
technique(s):
direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1F11, monoclonal

mol wt

60 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

Immunogen

Purified recombinant fragment of human SRC expressed in E.coli.
Mouse monoclonal antibody raised against SRC

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angeles Alvarez Secord et al.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society, 24(2), 218-225 (2014-01-11)
This study aimed to explore the activity of dasatinib in combination with docetaxel, gemcitabine, topotecan, and doxorubicin in ovarian cancer cells. Cells with previously determined SRC pathway and protein expression (SRC pathway/SRC protein IGROV1, both high; SKOV3, both low) were
M Ferrandi et al.
Human molecular genetics, 23(16), 4371-4382 (2014-04-05)
Mutant α-adducin and endogenous ouabain levels exert a causal role in hypertension by affecting renal Na-K ATPase. In addition, mutant β-adducin is involved in glomerular damage through nephrin down-regulation. Recently, the salt-inducible kinase 1 (SIK1) has been shown to exert
Mattia Capulli et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 29(8), 1833-1846 (2014-03-13)
Chondroadherin (CHAD) is a leucine-rich protein promoting cell attachment through binding to integrin α2 β1 and syndecans. We observed that CHAD mRNA and protein were lower in bone biopsies of 50-year-old to 65-year-old osteoporotic women and in bone samples of
Steven J McElroy et al.
The American journal of pathology, 184(10), 2768-2778 (2014-09-14)
Necrotizing enterocolitis (NEC) affects up to 10% of premature infants, has a mortality of 30%, and can leave surviving patients with significant morbidity. Neuregulin-4 (NRG4) is an ErbB4-specific ligand that promotes epithelial cell survival. Thus, this pathway could be protective
Duangrat Tantikanlayaporn et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(8-9), 676-682 (2013-04-06)
Curcuma comosa Roxb. is ginger-family plant used to relieve menopausal symptoms. Previous work showed that C. comosa extracts protect mice from ovariectomy-induced osteopenia with minimal effects on reproductive organs, and identified the diarylheptanoid (3R)-1,7-diphenyl-(4E,6E)-4,6-heptadien-3-ol (DPHD) as the major active component

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service