Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4702135

Monoclonal Anti-CD3 PerCP produced in mouse

new

Synonym(s):

CD3E, T3E, TCRE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

Peridinin-Chlorophyll-Protein Complex conjugate

Quality Level

antibody product type

primary antibodies

clone

SK7, monoclonal

form

buffered aqueous solution

species reactivity

( Human), (Non-human primates)

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD3E(916)

General description

CD3 complex is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkynje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases.

Specificity

The mouse monoclonal antibody SK7 recognizes an extracellular epitope on the CD3 antigen on mature human T cells. This antibody reacts with the epsilon chain of the CD3 complex. The monoclonal antibodies SK7 and UCHT1 recognize overlapping epitopes.

Immunogen

Human thymocytes

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated Peridinin-Chlorophyll Protein (PerCP) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service