Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700591

Sigma-Aldrich

Monoclonal Anti-CD62L, low endotoxin antibody produced in rat

clone MEL-14, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-L-selectin, Anti-Sell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEL-14, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Sell(20343)

Related Categories

General description

The rat monoclonal antibody MEL-14 reacts with mouse CD62L (L-selectin), a 75 kDa single chain type I glycoprotein expressed on most peripheral blood B lymphocytes, T lymphocytes, monocytes and granulocytes; it is also present on a subset of NK cells and certain hematopoietic malignant cells.

Immunogen

C3H/eb mouse B cell lymphoma 38C-13

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 0.5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 10 EU/mg of the protein, as determined by the LAL test.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heping Xu et al.
Blood, 112(4), 1166-1174 (2008-04-09)
Using noninvasive in vivo imaging and experimental autoimmune uveoretinitis as a model, we show for the first time that the mechanisms controlling blood monocyte recirculation through peripheral and lymphoid tissues alter during inflammation. The recirculation of monocytes in mice with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service