Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500378

Sigma-Aldrich

Anti-Collagen IV α3 (Cleaved-Pro1426) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CO4A3, COL4A3, Collagen α-3(IV) chain, Goodpasture antigen, Tumstatin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Pro1426)

Gene Information

human ... COL4A3(1285)

General description

Collagen type IV alpha 3 chain (COL4A3) gene with 52 exons, codes for the α3 chain of type IV collagen. COL4A3 protein is the vital structural protein in the basement membrane. COL4A3 gene is located on human chromosome 2q36.3. Anti-Collagen IV α3 (cleaved-pro1426) antibody detects endogenous levels of fragment of activated collagen IV α3 (cleaved-pro1426) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Collagen IV alpha3.

Immunogen Range: 1407-1456

Biochem/physiol Actions

Type IV collagen plays a key role in cell adhesion, proliferation, migration, survival, and differentiation. It maintains the assembly and mechanical stability by acting as a scaffold. Collagen type IV alpha 3 chain (COL4A3) mutations are associated with Alport syndrome (AS). Heterozygous mutations in the COL4A4 gene are observed in individuals with benign familial hematuria (BFH). Alterations in the COL4A3 gene also result in diabetic kidney disease from maturity-onset diabetes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Cervera-Acedo et al.
BMC nephrology, 18(1), 325-325 (2017-11-02)
Alport syndrome is an inherited renal disorder characterized by glomerular basement membrane lesions with hematuria, proteinuria and frequent hearing defects and ocular abnormalities. The disease is associated with mutations in genes encoding α3, α4, or α5 chains of type IV
Laurence Heidet et al.
Journal of the American Society of Nephrology : JASN, 12(1), 97-106 (2001-01-03)
Mutations in either the COL4A3 or the COL4A4 genes, encoding the alpha3 and alpha4 chains of type IV collagen, are responsible for the autosomal-recessive form of Alport syndrome, a progressive hematuric nephropathy characterized by glomerular basement membrane abnormalities. Reported here
Liang Xia et al.
Disease markers, 2019, 8705989-8705989 (2019-03-19)
Mutations in the COL4A3 gene are frequently reported to be associated with various types of hereditary nephropathy. COL4A3 encodes the α3 chain of type IV collagen, which is the main structural protein in the basement membrane. Mutations in this gene
Daniel Öhlund et al.
BMC cancer, 13, 154-154 (2013-03-28)
Pancreatic cancer shows a highly aggressive and infiltrative growth pattern and is characterized by an abundant tumor stroma known to interact with the cancer cells, and to influence tumor growth and drug resistance. Cancer cells actively take part in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service