Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4301846

Sigma-Aldrich

Anti-CXCL6

affinity isolated antibody

Synonym(s):

CKA-3, GCP-2, SCYB6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1: 100-300

accession no.

NP_002984

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCL6(6372)

General description

CXCL6 (C-X-C motif chemokine ligand 6)/GCP-2 (granulocyte chemotactic protein-2), a pro-inflammatory neutrophil chemoattractant, is mapped to human chromosome 4q12-q13. It belongs to the ELR+ CXC family. It is synthesized by mesenchymal cells like fibroblasts and micro and macrovascular endothelial cells.

Immunogen

Synthetic peptide of human CXCL6

Biochem/physiol Actions

CXCL6 (C-X-C motif chemokine ligand 6)/GCP-2 (granulocyte chemotactic protein-2) plays an important role in the recruitment and activation of neutrophils. It acts as an autocrine growth factor resulting in tumor progression and metastasis. CXCL6 participates in menstruation, implantation and the maintenance of early pregnancy. GCP-2 plays an important role in mucosal chronic inflammatory conditions like inflammatory bowel disease or chronic rhinosinusitis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pooja Mittal et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 60(3), 246-257 (2008-09-11)
CXCL6 is a potent pro-inflammatory neutrophil chemoattractant and activator whose activity during pregnancy is not well-established. The purpose of this study was to determine if CXCL6 is present in amniotic fluid (AF) and if CXCL6 concentrations in AF change with
M Kebschull et al.
Journal of periodontal research, 44(4), 465-471 (2008-10-10)
Mucosal inflammatory responses are orchestrated largely by pro-inflammatory chemokines. The chemokine granulocyte chemotactic protein 2 (CXCL6) is involved in neutrophil recruitment and migration. Previous studies have shown that granulocyte chemotactic protein 2 is up-regulated during mucosal inflammation (e.g. in inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service