Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300526

Sigma-Aldrich

Anti-ITGB3 (Ab-785) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD61 antibody produced in rabbit, Anti-GP3A antibody produced in rabbit, Anti-GPIIIa antibody produced in rabbit, Anti-integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~110 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(I-T-Y-R-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGB3(3690)

Immunogen

Peptide sequence around aa. 783-787 (I-T-Y-R-G), according to the protein ITGB3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The Itg-Beta3 gene encodes the beta subunit of the platelet membrane adhesive protein receptor complex Itg-Beta3 protein, which belongs to the Integrin class of cell adhesion molecule receptors that share a common heterotrimeric structure with Alpha and Beta subunits. It is a common Beta subunit of the platelet complex and of two other Integrins (Fibronectin and Vitronectin Receptors), which have distinct Alpha subunits. Itg-Beta3 has been implicated in a wide variety of functions, including platelet aggregation and thrombosis and implantation, placentation, angiogenesis, bone remodeling, and tumor progression. Glanzmann Thrombasthenia can result from defects in the genes for either the Itg-Alpha2B or the Itg-Beta3 subunit.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Badrnya et al.
Thrombosis and haemostasis, 112(1), 128-136 (2014-02-28)
Circulating plasma microvesicles (PMVs) and their microRNA content are involved in the development of atherosclerosis and could serve as biomarkers for cardiovascular disease (CVD) progression. However, little is known on how smoking influences the levels of PMVs and microRNA signatures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service