Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300138

Sigma-Aldrich

Anti-phospho-GRIN2B (pTyr1474) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC142178 antibody produced in rabbit, Anti-MGC142180 antibody produced in rabbit, Anti-NMDAR2B antibody produced in rabbit, Anti-NR2B antibody produced in rabbit, Anti-glutamate receptor, ionotropic, N-methyl D-aspartate 2B antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~190 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(H-V-YP-E-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1474)

Gene Information

human ... GRIN2B(2904)

Immunogen

Peptide sequence around phosphorylation site of Tyr1474 (H-V-Y(p)-E-K), according to the protein GRIN2B.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service