Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200580

Sigma-Aldrich

Anti-Msi-1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

AntiMSI1, AntiMSI1H, AntiMUSASHI 1, Antim-Msi-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~39 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using Lysates of HEK-293T cells overexpressing human Msi-1.
immunoprecipitation (IP): 5-10 μg using HEK-293T cells overexpressing human Msi-1.
indirect immunofluorescence: 5-10 μg/mL using HeLa cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSI1(4440)
rat ... Msi1(259272)

General description

Musashi-1 (Msi-1) is highly expressed in neural stem/progenitor cells (NS/PCs). MSI1 gene is mapped to human chromosome 12q24.

Immunogen

synthetic peptide corresponding to an internal sequence of human Msi-1, conjugated to KLH. The corresponding sequence is identical in rat and highly conserved (94% identity) in mouse Msi-1.

Application

Anti-Msi-1 antibody produced in rabbit has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Musashi-1 (Msi-1) is a neural RNA-binding protein that regulate translation of target mRNAs. It is associated with maintenance and asymmetric cell division of neural stem cells. Msi-1 binds to the 3′-untranslated region (UTR) of its target mRNAs in neural stem/progenitor cells (NS/PCs), repressing their translation, and interfering with NS/PC differentiation thus maintaining NP/PCs in the undifferentiated state. Loss of Msi-1 function disrupts the balance between germ-line stem cell renewal and differentiation, causing premature differentiation. Msi-1 has been shown to regulate the Notch and Wnt signaling pathway. Upregulation of Msi-1 augments Notch signaling, via translational repression of the Notch signaling antagonist m-Numb, thereby inhibiting the differentiation of neural stem cells into neurons. Msi-1 is also expressed in the nucleus during the early neural differentiation of mouse embryonic stem cells (ESCs) and found to act synergistically with Lin28 as a novel cofactor for the blockade of let-7 family miRNA biogenesis. Expression of Msi1- has been detected in several human cancers including brain tumors, suggesting its role in oncogenic development.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Musashi1 cooperates in abnormal cell lineage protein 28 (Lin28)-mediated let-7 family microRNA biogenesis in early neural differentiation
Kawahara H, et al.
The Journal of Biological Chemistry, 286(18), 16121-16130 (2011)
Musashi1: an evolutionally conserved marker for CNS progenitor cells including neural stem cells
Kaneko Y, et al.
Developmental Neuroscience, 22(1-2), 139-139 (2000)
The RNA-binding protein Musashi1 affects medulloblastoma growth via a network of cancer-related genes and is an indicator of poor prognosis
Vo DT, et al.
The American Journal of Pathology, 181(5), 1762-1772 (2012)
RNA-binding protein Musashi family: roles for CNS stem cells and a subpopulation of ependymal cells revealed by targeted disruption and antisense ablation
Sakakibara S, et al.
Proceedings of the National Academy of Sciences of the USA, 99(23), 15194-15199 (2002)
The neural RNA-binding protein Musashi1 translationally regulates mammalian numb gene expression by interacting with its mRNA
Imai T, et al.
Molecular and Cellular Biology, 21(12), 3888-3900 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service