Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200562

Sigma-Aldrich

Anti-AOX1 antibody, Mouse monoclonal

clone AO15, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-AO, Monoclonal Anti-AOH1, Monoclonal Anti-AOX1 antibody produced in mouse, Monoclonal Anti-EC 1.2.3.1, Monoclonal Anti-aldehyde oxidase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

AO15, monoclonal

form

buffered aqueous solution

mol wt

antigen ~148 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1.0-2.0 μg/mL using extracts of HepG2 cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AOX1(316)

General description

Monoclonal Anti-AOX1 (mouse IgG1 isotype) is derived from the hybridoma AO15 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Aldehyde oxidase 1 (AOX1) comprises the 2Fe/2S clusters and two flavin adenine nucleotide (FAD) domain-containing N-terminal domain I and II, respectively. The C-terminal domain encompasses domain III with a substrate-binding site. The gene encoding AOX1 is mapped to human chromosome 2q33.1.

Specificity

Monoclonal Anti-AOX1 recognizes human AOX1.

Immunogen

synthetic peptide corresponding to an internal sequence of human AOX1. The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-AOX1 antibody produced in mouse has been used in reverse-phase protein array analysis(5) and immunoblotting.

Biochem/physiol Actions

Aldehyde oxidase 1 (AOX1), along with the endoplasmic cytochrome P450 system (CYP450), participates in the drug metabolism and generation of reactive oxygen species. It also has a role in the hepatic phase I metabolism of numerous xenobiotics. Reduced AOX1 protein expression is implicated in chronic pancreatitis, and hepatocellular carcinoma. Furthermore, hypermethylation of the AOX1 promoter region is observed in colon cancer and prostate cancer, making it a potential biomarker for prostate cancer diagnosis. Polymorphism in the AOX1 gene also displays variation in metabolism of the quinoxaline phenoxypropionic acid derivative XK469.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,isnot recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A pharmacogenetic study of aldehyde oxidase I in patients treated with XK469
Ramirez J, et al.
Pharmacogenetics and Genomics, 24(2), 129-129 (2014)
Kalpana D Acharya et al.
eNeuro, 4(5) (2017-09-29)
Progestins bind to the progestin receptor (PR) isoforms, PR-A and PR-B, in brain to influence development, female reproduction, anxiety, and stress. Hormone-activated PRs associate with multiple proteins to form functional complexes. In the present study, proteins from female mouse hypothalamus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service