Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200470

Sigma-Aldrich

Anti-SND1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Staphylococcal nuclease and tudor domain containing 1, p100, Anti-TDRD11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~102 kDa

species reactivity

rat, human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using lysates of rat NRK cells.
indirect immunofluorescence: 2.5-5.0 μg/mL using human HeLa cells.
western blot: 0.25-0.5 μg/mL using whole extracts of mouse NIH-3T3 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SND1(27044)
mouse ... snd1(56463)
rat ... snd1(64635)

General description

Staphylococcal nuclease and tudor domain containing 1 (SND1), also called as Tudor-SN, is encoded by the gene mapped to human chromosome 7q32.1. The encoded protein acts as an important constituent of RNA-induced silencing complex (RISC). SND1 is characterized with an five repeated staphylococcal nuclease homology domains and a Tudor-homology domain involved in interaction of nucleic acids and proteins, respectively.
The SND1 protein is a part of the RNA-induced silencing complex (RISC). It is highly conserved from yeast to human.

Specificity

Anti-SND1 recognizes human, rat and mouse SND1.

Immunogen

peptide corresponding to an internal region of human SND1, conjugated to KLH. The corresponding sequence differs by a single amino acid in mouse and rat.

Application

Anti-SND1 antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

SND1 protein plays a vital role in the degradation of hyper-edited inosine containing miRNA precursors. It also controls miRNA processing and expression through RNA editing by adenosine deaminase acting on RNA (ADAR).
Staphylococcal nuclease and tudor domain containing 1 (SND1) interacts with RNA in some sequence-specific manner and regulates the gene expression at both transcriptional and posttranscriptional levels. The encoded protein plays a key role in gene transcription and mRNA metabolism. Additionally, it is also implicated in maintenance of cellular cholesterol distribution and homeostasis in hepatoma cells. SND1 facilitates various RNA functions such as, RNA maturation, spliceosome assembly,pre-mRNA splicing, RNA stability and RNA editing. It is also involved in RNA interference,microRNA processing, response to environmental and metabolic stressors and lipoprotein lipid secretion. Overexpression of the gene has been observed in human colon cancers and hepatocellular carcinoma cells.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SND1, a Component of RNA-Induced Silencing Complex, Is Up-regulated in Human Colon Cancers and Implicated in Early Stage Colon Carcinogenesis
Tsuchiya N
Cancer Research, 67, 9568-9576 (2007)
SND1 overexpression deregulates cholesterol homeostasis in hepatocellular carcinoma.
Navarro-Imaz H
Biochimica et Biophysica Acta, 1861, 988-996 (2016)
Prognostic implication of MET overexpression in myxofibrosarcomas: an integrative array comparative genomic hybridization, real-time quantitative PCR, immunoblotting, and immunohistochemical analysis.
Lee JC
Modern Pathology, 23, 1379-1392 (2010)
Expression analysis of Tudor-SN protein in mouse tissues
Fashe T, et al.
Tissue & cell, 45(1), 21-31 (2013)
Enara Arretxe et al.
Nucleic acids research, 43(22), 10673-10688 (2015-09-02)
The NF-κB-inducible Staphylococcal nuclease and tudor domain-containing 1 gene (SND1) encodes a coactivator involved in inflammatory responses and tumorigenesis. While SND1 is known to interact with certain transcription factors and activate client gene expression, no comprehensive mapping of SND1 target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service