Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200461

Sigma-Aldrich

Anti-Arc (N-terminal) antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Activity-regulated cytoskeleton-associated protein, Anti-Arc3.1, Anti-Arg3.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

rat, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 20-30 μg/mL using formalin-fixed, paraffin-embedded rat brain.
immunoprecipitation (IP): 2-4 μg using lysates of HEK-293T cells over-expressing human Arc.
western blot: 1.5-3.0 μg/mL using extracts of HEK-293T cells over-expressing human Arc.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARC(23237)
rat ... Arc(54323)

General description

Activity-regulated cytoskeleton-associated protein (Arc) is a growth factor and immediate-early gene (IEG) that is enriched in the brain. Arc is located in the distal dendrites and spine. The Arc gene is located on the human chromosome at 8q24.3.

Specificity

Anti-Arc(N-terminal) specifically recognizes humanand rat Arc.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal region of human Arc, conjugated to KLH. The corresponding sequence is highly conserved (90% sequence identity) in rat and mouse Arc.

Application

Anti-Arc (N-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunohistochemistry

Biochem/physiol Actions

Activity-regulated cytoskeleton-associated protein (Arc) induction is required for late long-term potentiation (LTP) and memory consolidation. It also plays a role in LTP-induced cofilin phosphorylation and F-actin stabilization. This protein affects synaptic plasticity. Arc regulates the endocytosis of α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid (AMPA) receptors through its interaction with endocytic proteins endophilin2/3 and dynamin. It is associated with notch signaling and spine morphology.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution bycentrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah A Stern et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(9), 2179-2190 (2014-03-20)
To treat cognitive disorders in humans, new effective therapies that can be easily delivered systemically are needed. Previous studies showed that a bilateral injection of insulin-like growth factor II (IGF-II) into the dorsal hippocampus of rats or mice enhances fear
Jayme R McReynolds et al.
Neurobiology of learning and memory, 115, 49-57 (2014-09-10)
The basolateral complex of the amygdala (BLA) plays a role in the modulation of emotional memory consolidation through its interactions with other brain regions. In rats, memory enhancing infusions of the β-adrenergic receptor agonist clenbuterol into the BLA immediately after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service