Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200330

Sigma-Aldrich

Anti-phospho-CD3ζ (pTyr72) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CD247, Anti-CD3H, Anti-CD3Q, Anti-CD3Z, Anti-T cell receptor T3 zeta chain, Anti-T3Z, Anti-TCRK, Anti-Tcrz

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunoprecipitation (IP): 2-4 μg using lysates of HEK-293T cells co-expressing mCD3ϲ and constitutively active LCK.
western blot: 1:1,000-1:2,000 using lysates of HEK-293T cells co-expressing mCD3ϲ and constitutively active LCK.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr72)

Gene Information

mouse ... Cd247(12503)

General description

T cell antigen receptor ζ chain (CD3ζ), also known as CD247 gene, spanning 88kb of genomic DNA, is mapped to human chromosome 1q24.2.
The T cell antigen receptor-CD3 (TCR-CD3) multimeric complex is composed of TCRαβ, CD3γε, CD3δε and ζ ζ dimers. The CD3γ, CD3δ and CD3ε, each contain one immunoreceptor tyrosine-based activation motif (ITAM), whereas the ζ chain contains three ITAMs.

Immunogen

synthetic phosphopeptide corresponding to residues surrounding Tyr72 of mouse CD3ζ, conjugated to KLH. The corresponding sequence differs by a single amino acid in human.

Application

Anti-phospho-CD3ζ (pTyr72) antibody has been used in
  • immunoblotting
  • immunoprecipitation
  • western blotting

Biochem/physiol Actions

T cell antigen receptor ζchain (CD3ζ) functions as a key signal transduction component of the T cell antigen receptor (TCR) complex. The encoded protein facilitates optimal effector T-cell function by enhancing receptor expression and signaling. Mutation in the gene increases the risk of susceptibility to systemic lupus erythematosus (SLE). Reduced expression of the gene has been observed in T-cells of cancer, lupus and chronic infectious diseases such as leprosy and tuberculosis patients. CD3ζ serves as a potent biomarker for determining progression and severity in patients with type 2 diabetes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crippling of CD3-zeta ITAMs does not impair T cell receptor signaling
Ardouin L, et al.
Immunity, 10(4), 409-420 (1999)
An immunoproteomic approach to characterize the CAR interactome and signalosome
Ramello M C, et al.
Science Signaling, 12(568), eaap9777-eaap9777 (2019)
Genetic Association of CD247 (CD3?) with SLE in a Large-Scale Multiethnic Study
Martins M
Genes and Immunity, 16, 142-150 (2015)
CD247, a Novel T Cell?Derived Diagnostic and Prognostic Biomarker for Detecting Disease Progression and Severity in Patients With Type 2 Diabetes
Eldor R
Diabetes Care, 38, 113-118 (2015)
Regulation of T Cell Receptor CD3? Chain Expression byL-Arginine
Rodriguez PC
The Journal of Biological Chemistry, 277, 21123-21129 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service