Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3701435

Sigma-Aldrich

Monoclonal Anti-Hepatitis Virus (Strain A59) Nonstructural Protein 9 (nsp9) antibody produced in mouse

clone 2C6.H1, purified immunoglobulin

Synonym(s):

Anti-Cas like docking antibody, Anti-nsp9 protein antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C6.H1, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG2bκ

shipped in

dry ice

storage temp.

−20°C

Gene Information

Escherichia coli ... NSP9(1489749)

Specificity

This antibody is directed against the MHV-A59 nsp9 protein. This product was purified from tissue culture supernatant fluid by Protein A chromatography.  No cross reactivity occurs with SARS CoV nsp9.  Cross reactivity with homologues from other sources has not been tested.

Immunogen

E.coli derived full-length MHV-A59 nsp9 protein.  This protein is part of the viral replicase polyprotein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% (w/v) Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service