Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3500910

Sigma-Aldrich

Anti-WEST NILE VIRUS MATRIX antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
clone:
polyclonal
species reactivity:
virus
citations:
3
technique(s):
ELISA: suitable

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

virus

concentration

1 mg/mL

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the West Nile virus matrix protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500910.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Wengler et al.
Journal of virology, 63(6), 2521-2526 (1989-06-01)
Flaviviruses are enveloped viruses which accumulate in cellular vacuoles prior to release. The membrane of cell-associated virus contains the proteins pre-M and E. During release of virus the pre-M protein is cleaved, and only its carboxy-terminal segment remains associated with
Justin Jang-Hann Chu et al.
The Journal of biological chemistry, 279(52), 54533-54541 (2004-10-12)
The functional receptor for the flavivirus West Nile (WNV) infection has been characterized in this study with a combination of biochemical and molecular approaches. A 105-kDa protease-sensitive glycoprotein that binds WNV was isolated from the plasma membrane of cells permissive
L Hannah Gould et al.
The Journal of clinical investigation, 113(8), 1102-1107 (2004-04-16)
West Nile virus was first detected in North America in 1999 and has subsequently spread throughout the United States and Canada and into Mexico and the Caribbean. This review describes the epidemiology and ecology of West Nile virus in North

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service