Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3500370

Sigma-Aldrich

Anti-TRBP1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TAR RNA binding protein 1, Anti-TARBP1, Anti-TRP-1, Anti-TRP-185

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
4
technique(s):
indirect ELISA: suitable
western blot: suitable

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARBP1(6894)

General description

The gene TARBP1 (TAR (HIV-1) RNA binding protein 1) is mapped to human chromosome 1q42.2. The protein has dsRBDs (double stranded RNA binding domains), a KR motif in the dsRBD2 and a Medipal domain.

Immunogen

TRBP1 antibody was raised against a 17 amino acid peptide from near the center of human TRBP1.

Biochem/physiol Actions

TARBP1 (TAR (HIV-1) RNA binding protein 1) binds strongly to the small hairpin trans-activation responsive (TAR) RNA of HIV (human immunodeficiency virus)-1. It might have a role in disengaging RNA polymerase II from HIV-1 TAR RNA. In addition, it is also a part of the RNA-induced silencing complex (RISC). It interacts with Dicer and helps in processing of pre-microRNAs. TARBP1 also participates in other cellular functions, such as protein synthesis, RNA post-translational modifications, cellular proliferation, gene expression and stress-associated cellular processes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500370.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert D Beech et al.
Alcoholism, clinical and experimental research, 38(11), 2743-2753 (2014-11-26)
Alterations in stress-related gene expression may play a role in stress-related drinking and the risk of alcohol dependence. Microarrays were used to measure changes in gene expression in peripheral blood in nonsmoking, social drinking subjects exposed to 3 types of
Orla M Keane et al.
BMC genomics, 7, 42-42 (2006-03-07)
Gastrointestinal nematodes constitute a major cause of morbidity and mortality in grazing ruminants. Individual animals or breeds, however, are known to differ in their resistance to infection. Gene expression profiling allows us to examine large numbers of transcripts simultaneously in
F Wu-Baer et al.
The EMBO journal, 14(23), 5995-6009 (1995-12-01)
Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element located downstream of the transcription initiation site known as TAR. To characterize cellular factors that bind to TAR RNA and are involved in the
Sandrine Cornaz-Buros et al.
Cancer research, 74(22), 6610-6622 (2014-09-28)
Plasticity in cancer stem-like cells (CSC) may provide a key basis for cancer heterogeneity and therapeutic response. In this study, we assessed the effect of combining a drug that abrogates CSC properties with standard-of-care therapy in a Ewing sarcoma family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service