Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900239

Sigma-Aldrich

Anti-KDR (AB2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Ly73, Quek1, fetal liver kinase 1, flk-1, kinase insert domain receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDR(3791)

Immunogen

Synthetic 16 amino acid peptide from internal region of human VEGFR2. Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%); Gibbon, Marmoset (94%); Mouse, Dog, Hamster, Elephant, Pig (88%); Rat, Bovine, Panda, Horse, Opossum, Turkey (81%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Stacchiotti et al.
European journal of cancer (Oxford, England : 1990), 50(17), 3021-3028 (2014-10-02)
To explore the activity of pazopanib in solitary fibrous tumour (SFT). In a preclinical study, we compared the activity of pazopanib, sorafenib, sunitinib, regorafenib, axitinib and bevacizumab in a dedifferentiated-SFT (DSFT) xenotransplanted into Severe Combined Immunodeficiency (SCID) mice. Antiangiogenics were
Dae-Young Park et al.
The Journal of clinical investigation, 124(9), 3960-3974 (2014-07-26)
Schlemm's canal (SC) is a specialized vascular structure in the eye that functions to drain aqueous humor from the intraocular chamber into systemic circulation. Dysfunction of SC has been proposed to underlie increased aqueous humor outflow (AHO) resistance, which leads
Weiyong Shen et al.
PloS one, 9(8), e104759-e104759 (2014-08-15)
Royal College of Surgeons (RCS) rats develop vasculopathy as photoreceptors degenerate. The aim of this study was to examine the effect of erythropoietin (EPO) on retinopathy in RCS rats. Fluorescein angiography was used to monitor retinal vascular changes over time.
Kusum Rathore et al.
BMC cancer, 14, 465-465 (2014-06-27)
Development and further characterization of animal models for human cancers is important for the improvement of cancer detection and therapy. Canine bladder cancer closely resembles human bladder cancer in many aspects. In this study, we isolated and characterized four primary
Yi Zhou et al.
Biotechnology journal, 9(7), 944-953 (2014-01-15)
Bone tissue deficiency is a common clinical challenge. Tissue-engineered bone constructs are an effective approach for the repair of orthopedic bone defects. Mimicking the essential components of the in vivo microenvironment is an efficient way to develop functional constructs. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service