Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB2702301

Sigma-Aldrich

Monoclonal Anti-UQCRC1 antibody produced in mouse

clone GT139, affinity isolated antibody

Synonym(s):

D3S3191, QCR1, UQCR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT139, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

concentration

1mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 500-3000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... UQCRC1(7384)

Immunogen

Recombinant protein encompassing a sequence within the center region of human UQCRC1.

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service