Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501501

Sigma-Aldrich

Anti-FOXC1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARA, Anti-FKHL7, Anti-FREAC3, Anti-HGNC:3800, Forkhead box C1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXC1(2296)

Immunogen

Peptide with sequence DAVKDKEEKDRLH, from the internal region of the protein sequence according to NP_001444.2

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Alvares et al.
The Journal of physiology, 592(14), 3089-3111 (2014-06-01)
PreBötzinger complex inspiratory rhythm-generating networks are excited by metabotropic purinergic receptor subtype 1 (P2Y1R) activation. Despite this, and the fact that inspiratory MNs express P2Y1Rs, the role of P2Y1Rs in modulating motor output is not known for any MN pool.
Li Wang et al.
The Journal of physiology, 592(16), 3559-3576 (2014-06-29)
Striatal dopamine (DA) is critically involved in major brain functions such as motor control and deficits such as Parkinson's disease. DA is released following stimulation by two pathways: the nigrostriatal pathway and the cholinergic interneuron (ChI) pathway. The timing of
Lais Takata Walter et al.
Experimental neurology, 261, 510-517 (2014-08-15)
In the nervous system within physiological conditions, nitric oxide (NO) production depends on the activity of nitric oxide synthases (NOSs), and particularly on the expression of the neuronal isoform (nNOS). In the sensory systems, the role of NO is poorly
Jacob Wienecke et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(36), 11984-12000 (2014-09-05)
Serotonin (5-HT), an important modulator of both sensory and motor functions in the mammalian spinal cord, originates mainly in the raphe nuclei of the brainstem. However, following complete transection of the spinal cord, small amounts of 5-HT remain detectable below
Irene E Whitney et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(30), 10109-10121 (2014-07-25)
The retina contains two populations of cholinergic amacrine cells, one positioned in the ganglion cell layer (GCL) and the other in the inner nuclear layer (INL), that together comprise ∼1/2 of a percent of all retinal neurons. The present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service