Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2500981

Sigma-Aldrich

Anti-SP1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HGNC:11205

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP1(6667)

General description

Sp1 protein was the first transcription factor to be cloned and characterized. It was first detected in HeLa cells on the basis of its ability to activate the SV40 early promoter transcription. Analysis of structure and function has revealed that Sp1 can be separated into discrete functional domains. The DNA-binding domain consists of three zinc fingers that specifically bind to the GC-box element.

Immunogen

Peptide with sequence CSRIESPNENSNNSQ from the internal region of the protein sequence according to NP_612482.2.

Biochem/physiol Actions

Sp1 protein was shown to recognize and bind selectively to a GC-rich consensus sequence (GC-box: GGGCGG or CACCC) that presents in the promoter of several important cellular genes, including Simian vacuolating virus 40 (SV40) early, human immunodeficiency virus-1 (HIV-1), platelet-derived growth factor subunit B (PDGF-B), Myc, c-Src etc. In addition to transcription, Sp1 function has been linked to cell growth, cancer and Huntington disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structures of zinc finger domains from transcription factor Sp1. Insights into sequence-specific protein-DNA recognition.
Narayan VA
The Journal of Biological Chemistry, 272(12), 7801-7809 (1997)
Decreased association of the transcription factor Sp1 with genes downregulated in Huntington's disease.
Chen-Plotkin AS
Neurobiology of Disease, 22(2), 233-241 (2006)
Mithramycin is a gene-selective Sp1 inhibitor that identifies a biological intersection between cancer and neurodegeneration.
Sleiman SF
The Journal of Neuroscience, 31(18), 6858-6870 (2011)
Deepti Malik et al.
Molecular cancer, 13, 175-175 (2014-07-20)
microRNAs (miRNAs) play both oncogenic and oncostatic roles in leukemia. However, the molecular details underlying miRNA-mediated regulation of their target genes in pediatric B- and T-cell acute lymphoblastic leukemias (ALLs) remain unclear. The present study investigated the relationship between miR-2909
Carolina Vizcaíno et al.
PloS one, 9(8), e104687-e104687 (2014-08-12)
Ovarian cancer has a poor prognosis due to intrinsic or acquired resistance to some cytotoxic drugs, raising the interest in new DNA-binding agents such as mithramycin analogues as potential chemotherapeutic agents in gynecological cancer. Using a genome-wide approach, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service