Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

SAB1411621

Sigma-Aldrich

Anti-CD247 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CD3-ZETA, CD3H, CD3Q, CD3Z, T3Z, TCRZ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 18.7 kDa

species reactivity

human

technique(s)

proximity ligation assay: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD247(919)

General description

T cell antigen receptor ζ chain (CD3 ζ), also known as cluster of differentiation 247 (CD247) gene, spanning 88 kb of genomic DNA, is mapped to human chromosome 1q24.2.
The protein encoded by this gene is T-cell receptor ζ, which together with T-cell receptor α/β and γ/δ heterodimers, and with CD3-γ, -δ and -ε, forms the T-cell receptor-CD3 complex. The ζ chain plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. Low expression of the antigen results in impaired immune response. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CD247 (NP_932170.1, 1 a.a. ~ 164 a.a) full-length human protein.

Sequence
MKWKALFTAAILQAQLPITEAQSFGLLDPKLCYLLDGILFIYGVILTALFLRVKFSRSADAPAYQQGQNQLYNELNLGRREEYDVLDKRRGRDPEMGGKPQRRKNPQEGLYNELQKDKMAEAYSEIGMKGERRRGKGHDGLYQGLSTATKDTYDALHMQALPPR

Biochem/physiol Actions

T cell antigen receptor ζ chain (CD3 ζ)/cluster of differentiation 247 (CD247) functions as a key signal transduction component of the T cell antigen receptor (TCR) complex. The encoded protein facilitates optimal effector T-cell function by enhancing receptor expression and signaling. Mutation in the gene increases the risk of susceptibility to systemic lupus erythematosus (SLE). Reduced expression of the gene has been observed in T-cells of cancer, lupus and chronic infectious diseases such as leprosy and tuberculosis patients. CD247 serves as a potent biomarker for determining progression and severity in patients with type 2 diabetes.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD247, a Novel T Cell?Derived Diagnostic and Prognostic Biomarker for Detecting Disease Progression and Severity in Patients With Type 2 Diabetes
Eldor R, et al.
Diabetes Care (2014)
Genetic Association of CD247 (CD3ζ) with SLE in a Large-Scale Multiethnic Study
Martins M, et al.
Genes and Immunity, 16(2), 142-142 (2015)
Regulation of T Cell Receptor CD3ζ Chain Expression byL-Arginine.
Rodriguez PC, et al.
The Journal of Biological Chemistry, 277(24), 21123-21129 (2002)
Venugopal Gudipati et al.
Nature immunology, 21(8), 848-856 (2020-07-08)
Rational design of chimeric antigen receptors (CARs) with optimized anticancer performance mandates detailed knowledge of how CARs engage tumor antigens and how antigen engagement triggers activation. We analyzed CAR-mediated antigen recognition via quantitative, single-molecule, live-cell imaging and found the sensitivity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service