Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411157

Sigma-Aldrich

Anti-TRIB3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

C20orf97, NIPK, SINK, SKIP3, TRB3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 39.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIB3(57761)

General description

The protein encoded by this gene is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1. (provided by RefSeq)

Immunogen

TRIB3 (NP_066981.2, 1 a.a. ~ 358 a.a) full-length human protein.

Sequence
MRATPLAAPAGSLSRKKRLELDDNLDTERPVQKRARSGPQPRLPPCLLPLSPPTAPDRATAVATASRLGPYVLLEPEEGGRAYQALHCPTGTEYTCKVYPVQEALAVLEPYARLPPHKHVARPTEVLAGTQLLYAFFTRTHGDMHSLVRSRHRIPEPEAAVLFRQMATALAHCHQHGLVLRDLKLCRFVFADRERKKLVLENLEDSCVLTGPDDSLWDKHACPAYVGPEILSSRASYSGKAADVWSLGVALFTMLAGHYPFQDSEPVLLFGKIRRGAYALPAGLSAPARCLVRCLLRREPAERLTATGILLHPWLRQDPMPLAPTRSHLWEAAQVVPDGLGLDEAREEEGDREVVLYG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela D Filiou et al.
Journal of psychiatric research, 58, 115-122 (2014-08-16)
No comprehensive metabolic profile of trait anxiety is to date available. To identify metabolic biosignatures for different anxiety states, we compared mice selectively inbred for ∼ 40 generations for high (HAB), normal (NAB) or low (LAB) anxiety-related behavior. Using a
Devora Champa et al.
Endocrine-related cancer, 21(5), 755-767 (2014-07-12)
Poorly differentiated tumors of the thyroid gland (PDTC) are generally characterized by a poor prognosis due to their resistance to available therapeutic approaches. The relative rarity of these tumors is a major obstacle to our understanding of the molecular mechanisms
Weiwei Wang et al.
Diabetes research and clinical practice, 106(1), 101-109 (2014-08-13)
Fibrosis is the final disorder of most chronic kidney disease including diabetic nephropathy (DN), but the mechanisms are not fully understood. The present study aims to determine whether TRB3 participates in fibrogenesis in DN. Type1 diabetes was induced in male
Gisela Campos et al.
Archives of toxicology, 88(6), 1267-1280 (2014-04-22)
Since xenobiotics enter the organism via the liver, hepatocytes must cope with numerous perturbations, including modifications of proteins leading to endoplasmic reticulum stress (ER-stress). This triggers a signaling pathway termed unfolded protein response (UPR) that aims to restore homeostasis or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service