Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1410517

Sigma-Aldrich

Anti-GCHFR antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

GFRP, HsT16933, MGC138467, MGC138469, P35

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 9.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCHFR(2644)

General description

GTP cyclohydrolase I feedback regulatory protein binds to and mediates tetrahydrobiopterin inhibition of GTP cyclohydrolase I. The regulatory protein, GCHFR, consists of a homodimer. It is postulated that GCHFR may play a role in regulating phenylalanine metabolism in the liver and in the production of biogenic amine neurotransmitters and nitric oxide. (provided by RefSeq)

Immunogen

GCHFR (NP_005249.1, 1 a.a. ~ 84 a.a) full-length human protein.

Sequence
MPYLLISTQIRMEVGPTMVGDEQSDPELMQHLGASKRRALGNNFYEYYVDDPPRIVLDKLERRGFRVLSMTGVGQTLVWCLHKE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojie Liu et al.
Brain research bulletin, 106, 9-16 (2014-04-29)
N-methyl-D-aspartate receptor (NMDAR) and metabotropic glutamate receptor 5 (mGluR5) play an important role in nociceptive processing and central sensitization. Our previous study showed that tyrosine phosphorylation of NMDAR subunit 2B (NR2B) at Tyr1472 in spinal dorsal horn contributes to the
María Florencia Ogara et al.
Biochimica et biophysica acta, 1843(7), 1309-1324 (2014-04-08)
DNA damage, which perturbs genomic stability, has been linked to cognitive decline in the aging human brain, and mutations in DNA repair genes have neurological implications. Several studies have suggested that DNA damage is also increased in brain disorders such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service