Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB1409592

Sigma-Aldrich

Anti-RAD51C antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC104277, RAD51L2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD51C(5889)

Related Categories

General description

This gene is a member of the RAD51 family of related genes, which encode strand-transfer proteins thought to be involved in recombinational repair of damaged DNA and in meiotic recombination. This gene product interacts with two other DNA repair proteins, encoded by RAD51B and XRCC3, but not with itself. The protein copurifies with XRCC3 protein in a complex, reflecting their endogenous association and suggesting a cooperative role during recombinational repair. This gene is one of four localized to a region of chromosome 17q23 where amplification occurs frequently in breast tumors. Overexpression of the four genes during amplification has been observed and suggests a possible role in tumor progression. Alternative splicing has been observed for this gene and two variants encoding different isoforms have been identified. (provided by RefSeq)

Immunogen

RAD51C (NP_478123.1, 1 a.a. ~ 376 a.a) full-length human protein.

Sequence
MRGKTFRFEMQRDLVSFPLSPAVRVKLVSAGFQTAEELLEVKPSELSKEVGISKAEALETLQIIRRECLTNKPRYAGTSESHKKCTALELLEQEHTQGFIITFCSALDDILGGGVPLMKTTEICGAPGVGKTQLCMQLAVDVQIPECFGGVAGEAVFIDTEGSFMVDRVVDLATACIQHLQLIAEKHKGEEHRKALEDFTLDNILSHIYYFRCRDYTELLAQVYLLPDFLSEHSKVRLVIVDGIAFPFRHDLDDLSLRTRLLNGLAQQMISLANNHRLAVILTNQMTTKIDRNQALLVPALGESWGHAATIRLIFHWDRKQRLATLYKSPSQKECTVLFQIKPQGFRDTVVTSACSLQTEGSLSTRKRSRDPEEEL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service