Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408885

Sigma-Aldrich

Anti-RAET1E antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

LETAL, MGC125308, MGC125309, RAET1E2, ULBP4, bA350J20.7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 30.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAET1E(135250)

General description

Members of the RAET1 family, such as RAET1E, are major histocompatibility complex (MHC) class I-related genes located within a 180-kb cluster on chromosome 6q24.2-q25.3. RAET1 proteins contain MHC class I-like alpha-1 and alpha-2 domains. RAET1E and RAET1G (MIM 609244) differ from the other RAET1 proteins (e.g., RAET1I, or ULBP1; MIM 605697) in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. (Radosavljevic et al., 2002 [PubMed 11827464]).[supplied by OMIM

Immunogen

RAET1E (AAI03695.1, 1 a.a. ~ 263 a.a) full-length human protein.

Sequence
MRRISLTSSPVRLLLFLLLLLIALEIMVGGHSLCFNFTIKSLSRPGQPWCEAQVFLNKNLFLQYNSDNNMVKPLGLLGKKVNATSTWGELTQTLGEVGRDLRMLLCDIKPQIKTSDPSTLQVEMFCQHEAERCTGASWQFTINGEKSLLFDAMNMTWTVINHEASKIKETWKKDRGLEKYFRKLSKGDCDHWLREFLGHWEAMPEPTVSPVNASDIHWSSSSLPDRWIILGAFILLLLMGIVLICVWWQNGEWQAGLWPLRTS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service