Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408602

Sigma-Aldrich

Anti-CD46, (N-terminal) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MCP, MGC26544, MIC10, TLX, TRA2.10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~42.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD46(4179)

Related Categories

General description

The protein encoded by this gene is a type I membrane protein and is a regulatory part of the complement system. The encoded protein has cofactor activity for inactivation of complement components C3b and C4b by serum factor I, which protects the host cell from damage by complement. In addition, the encoded protein can act as a receptor for the Edmonston strain of measles virus, human herpesvirus-6, and type IV pili of pathogenic Neisseria. Finally, the protein encoded by this gene may be involved in the fusion of the spermatozoa with the oocyte during fertilization. This gene is found in a cluster on chromosome 1q32 with other genes encoding structural components of the complement system. At least fourteen different transcript variants encoding fourteen different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CD46 (NP_722548.1, 1 a.a. ~ 384 a.a) full-length human protein.

Sequence
MEPPGRRECPFPSWRFPGLLLAAMVLLLYSFSDACEEPPTFEAMELIGKPKPYYEIGERVDYKCKKGYFYIPPLATHTICDRNHTWLPVSDDACYRETCPYIRDPLNGQAVPANGTYEFGYQMHFICNEGYYLIGEEILYCELKGSVAIWSGKPPICEKVLCTPPPKIKNGKHTFSEVEVFEYLDAVTYSCDPAPGPDPFSLIGESTIYCGDNSVWSRAAPECKVVKCRFPVVENGKQISGFGKKFYYKATVMFECDKGFYLDGSDTIVCDSNSTWDPPVPKCLKVSTSSTTKSPASSASGPRPTYKPPVSNYPGYPKPEEGILDSLDVWVIAVIVIAIVVGVAVICVVPYRYLQRRKKKGKADGGAEYATYQTKSTTPAEQRG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Higgins et al.
Mediators of inflammation, 2014, 293925-293925 (2014-10-03)
CCL2 is an important inflammatory chemokine involved in monocyte recruitment to inflamed tissues. The extracellular nucleotide signalling molecules UTP and ATP acting via the P2Y2 receptor are known to induce CCL2 secretion in macrophages. We confirmed this in the human
Sibel P Yentur et al.
Neuropediatrics, 45(5), 309-313 (2014-05-31)
Subacute sclerosing panencephalitis (SSPE) is a late complication of measles infection. Immune dysfunction related to genetic susceptibility has been considered in disease pathogenesis. A functional single nucleotide polymorphism (SNP) of granzyme B gene (GZMB) reported in several pathologies may also
Jae-Won Lee et al.
Molecular and cellular biochemistry, 397(1-2), 109-116 (2014-08-06)
We previously demonstrated that 3,4,5-trihydorxycinnamic acid (THC), a derivative of hydroxycinnamic acids, possesses protective effect in lipopolysaccharide (LPS)-induced endotoxemia models. However, the effects of THC in LPS-induced septic kidney are still unclear. Therefore, the present study was carried out to
Lei Zhang et al.
Laboratory investigation; a journal of technical methods and pathology, 94(8), 839-850 (2014-05-28)
Peritoneal fibrosis is a complication of patients with long-term continuous ambulatory peritoneal dialysis (CAPD). Reports have indicated that angiotensin (Ang) II may correlate with the development of peritoneal fibrosis. However, it is unknown whether aldosterone also has a role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service