Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1407122

Sigma-Aldrich

Anti-SEC63 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ERdj2, PRO2507, SEC63L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
1
technique(s):
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~88 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEC63(11231)

Related Categories

General description

The Sec61 complex is the central component of the protein translocation apparatus of the endoplasmic reticulum (ER) membrane. The protein encoded by this gene and SEC62 protein are found to be associated with ribosome-free SEC61 complex. It is speculated that Sec61-Sec62-Sec63 may perform post-translational protein translocation into the ER. The Sec61-Sec62-Sec63 complex might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway. The encoded protein is an integral membrane protein located in the rough ER. (provided by RefSeq)

Immunogen

SEC63 (NP_009145.1, 1 a.a. ~ 760 a.a) full-length human protein.

Sequence
MAGQQFQYDDSGNTFFYFLTSFVGLIVIPATYYLWPRDQNAEQIRLKNIRKVYGRCMWYRLRLLKPQPNIIPTVKKIVLLAGWALFLFLAYKVSKTDREYQEYNPYEVLNLDPGATVAEIKKQYRLLSLKYHPDKGGDEVMFMRIAKAYAALTDEESRKNWEEFGNPDGPQATSFGIALPAWIVDQKNSILVLLVYGLAFMVILPVVVGSWWYRSIRYSGDQILIRTTQIYTYFVYKTRNMDMKRLIMVLAGASEFDPQYNKDATSRPTDNILIPQLIREIGSINLKKNEPPLTCPYSLKARVLLLSHLARMKIPETLEEDQQFMLKKCPALLQEMVNVICQLIVMARNREEREFRAPTLASLENCMKLSQMAVQGLQQFKSPLLQLPHIEEDNLRRVSNHKKYKIKTIQDLVSLKESDRHTLLHFLEDEKYEEVMAVLGSFPYVTMDIKSQVLDDEDSNNITVGSLVTVLVKLTRQTMAEVFEKEQSICAAEEQPAEDGQGETNKNRTKGGWQQKSKGPKKTAKSKKKKPLKKKPTPVLLPQSKQQKQKQANGVVGNEAAVKEDEEEVSDKGSDSEEEETNRDSQSEKDDGSDRDSDREQDEKQNKDDEAEWQELQQSIQRKERALLETKSKITHPVYSLYFPEEKQEWWWLYIADRKEQTLISMPYHVCTLKDTEEVELKFPAPGKPGNYQYTVFLRSDSYMGLDQIKPLKLEVHEAKPVPENHPQWDTAIEGDEDQEDSEGFEDSFEEEEEEEEDDD

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric M Davis et al.
Cell reports, 11(11), 1727-1736 (2015-06-16)
Glycophosphatidylinositol-anchored proteins (GPI-APs) play essential roles in physiology, but their biogenesis and trafficking have not been systematically characterized. Here, we took advantage of the recently available haploid genetics approach to dissect GPI-AP pathways in human cells using prion protein (PrP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service