Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1403345

Sigma-Aldrich

Monoclonal Anti-KIF9, (C-terminal) antibody produced in mouse

clone 4E9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC104186

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIF9(64147)

General description

KIF9 (Kinesin family member 9) is a microtubule-associated protein belonging to the kinesin 9 motor family. It is localized in the mitotic spindle.
Mouse monoclonal antibody raised against a partial recombinant KIF9.

Immunogen

KIF9 (NP_878905.1, 691 a.a. ~ 789 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VDQCRHRLLMEFDIWYNESFVIPEDMQMALKPGGSIRPGMVPVNRIVSLGEDDQDKFSQLQQRVLPEGPDSISFYNAKVKIEQKHNYLKTMMGLQQAHR

Application

Monoclonal Anti-KIF9, (C-terminal) antibody produced in mouse is suitable for western blot and indirect ELISA.

Biochem/physiol Actions

KIF9 (Kinesin family member 9) is involved in the regulation of spindle dynamics In terms of chromosome organization, spindle length control, and mitotic progression. It forms a complex by binding to the GEM, a small GTP-binding protein, in mitotic cells and the complex controls spindle length. It also retards microtubule growth. In primary macrophage of humans, KIF9 controls morphology and numbers of the podosomes and its ability to degrade matrix material.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susanne Cornfine et al.
Molecular biology of the cell, 22(2), 202-215 (2010-12-02)
Podosomes are actin-based matrix contacts in a variety of cell types, most notably monocytic cells, and are characterized by their ability to lyse extracellular matrix material. Besides their dependence on actin regulation, podosomes are also influenced by microtubules and microtubule-dependent
Guillaume Andrieu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(12), 5025-5034 (2012-09-12)
Within the Ras superfamily, Gem is a small GTP-binding protein that plays a role in regulating Ca(2+) channels and cytoskeletal remodeling in interphase cells. Here, we report for the first time that Gem is a spindle-associated protein and is required

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service