Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1402685

Sigma-Aldrich

Monoclonal Anti-ALCAM antibody produced in mouse

clone 1H3, ascites fluid

Synonym(s):

CD166, FLJ38514, MEMD, MGC71733

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H3, monoclonal

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALCAM(214)

Immunogen

ALCAM (NP_001618, 28 a.a. ~ 137 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
WYTVNSAYGDTIIIPCRLDVPQNLMFGKWKYEKPDGSPVFIAFRSSTKKSVQYDDVPEYKDRLNLSENYTLSISNARISDEKRFVCMLVTEDNVFEAPTIVKVFKQPSKP

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hung-Li Tsai et al.
PloS one, 9(8), e104937-e104937 (2014-08-30)
Wnts were previously shown to regulate the neurogenesis of neural stem or progenitor cells. Here, we explored the underlying molecular mechanisms through which Wnt signaling regulates neurotrophins (NTs) in the NT-induced neuronal differentiation of human mesenchymal stem cells (hMSCs). NTs
Y Chu et al.
Cancer gene therapy, 21(5), 200-208 (2014-05-24)
Anti-angiogenesis gene therapy is considered a promising treatment for excessive vascularization. Mesenchymal stem cell (MSC)-based gene therapy may enhance the effect of anti-angiogenesis by maintaining a long therapeutic period in vivo. However, transduction efficiencies and transgene expression in MSC-based gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service