Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1400590

Sigma-Aldrich

Anti-BCOR antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ANOP2, Anti-FLJ20285, Anti-KIAA1575, Anti-MAA2, Anti-MGC71031

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCOR(54880)

General description

The protein encoded by this gene was identified as an interacting corepressor of BCL6, a POZ/zinc finger transcription repressor that is required for germinal center formation and may influence apoptosis. This protein selectively interacts with the POZ domain of BCL6, but not with eight other POZ proteins. Specific class I and II histone deacetylases (HDACs) have been shown to interact with this protein, which suggests a possible link between the two classes of HDACs. Several transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

BCOR (NP_065977.1, 1 a.a. ~ 1004 a.a) full-length human protein.

Sequence
MLSATPLYGNVHSWMNSERVRMCGASEDRKILVNDGDASKARLELREENPLNHNVVDASTAHRIDGLAALSMDRTGLIREGLRVPGNIVYSSLCGLGSEKGREAATSTLGGLGFSSERNPEMQFKPNTPETVEASAVSGKPPNGFSAIYKTPPGIQKSAVATAEALGLDRPASDKQSPLNINGASYLRLPWVNPYMEGATPAIYPFLDSPNKYSLNMYKALLPQQSYSLAQPLYSPVCTNGERFLYLPPPHYVGPHIPSSLASPMRLSTPSASPAIPPLVHCADKSLPWKMGVSPGNPVDSHAYPHIQNSKQPRVPSAKAVTSGLPGDTALLLPPSPRPSPRVHLPTQPAADTYSEFHKHYARISTSPSVALSKPYMTVSSEFPAARLSNGKYPKAPEGGEGAQPVPGHARKTAVQDRKDGSSPPLLEKQTVTKDVTDKPLDLSSKVVDVDASKADHMKKMAPTVLVHSRAGSGLVLSGSEIPKETLSPPGNGCAIYRSEIISTAPSSWVVPGPSPNEENNGKSMSLKNKALDWAIPQQRSSSCPRMGGTDAVITNVSGSVSSAGRPASASPAPNANADGTKTSRSSVETTPSVIQHVGQPPATPAKHSSSTSSKGAKASNPEPSFKANENGLPPSSIFLSPNEAFRSPPIPYPRSYLPYPAPEGIAVSPLSLHGKGPVYPHPVLLPNGSLFPGHLAPKPGLPYGLPTGRPEFVTYQDALGLGMVHPMLIPHTPIEITKEEKPERRSRSHERARYEDPTLRNRFSEILETSSTKLHPDVPTDKNLKPNPNWNQGKTVVKSDKLVYVDLLREEPDAKTDTNVSKPSFAAESVGQSAEPPKPSVEPALQQHRDFIALREELGRISDFHETYTFKQPVFTVSKDSVLAGTNKENLGLPVSTPFLEPPLGSDGPAVTFGKTQEDPKPFCVGSAPPSVDVTPTYTKDGADEAESNDGKVLKPKPSKLAKRIANSAGYVGDRFKCVTTELYADSSQLSREQRALQVSPPT

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service