Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1400339

Sigma-Aldrich

Anti-PIWIL1 antibody produced in mouse

IgG fraction of antiserum

Synonym(s):

Anti-HIWI, Anti-PIWI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIWIL1(9271)

General description

This gene encodes a member of the PIWI subfamily of Argonaute proteins, evolutionarily conserved proteins containing both PAZ and Piwi motifs that play important roles in stem cell self-renewal, RNA silencing, and translational regulation in diverse organisms. The encoded protein may play a role as an intrinsic regulator of the self-renewal capacity of germline and hematopoietic stem cells. (provided by RefSeq)

Immunogen

PIWIL1 (NP_004755, 1 a.a. ~ 861 a.a) full-length human protein.

Sequence
MTGRARARARGRARGQETAQLVGSTASQQPGYIQPRPQPPPAEGELFGRGRQRGTAGGTAKSQGLQISAGFQELSLAERGGRRRDFHDLGVNTRQNLDHVKESKTGSSGIIVRLSTNHFRLTSRPQWALYQYHIDYNPLMEARRLRSALLFQHEDLIGKCHAFDGTILFLPKRLQQKVTEVFSKTRNGEDVRITITLTNELPPTSPTCLQFYNIIFRRLLKIMNLQQIGRNYYNPNDPIDIPSHRLVIWPGFTTSILQYENSIMLCTDVSHKVLRSETVLDFMFNFYHQTEEHKFQEQVSKELIGLVVLTKYNNKTYRVDDIDWDQNPKSTFKKADGSEVSFLEYYRKQYNQKITDLKQPVLVSQPKRRRGPGGTLPGPAMLIPELCYLTGLTDKMRNDFNVMKDLAVHTRLTPEQRQREVGRLIDYIHKNDNVQRELRDWGLSFDSNLLSFSGRILQTEKIHQGGKTFDYNPQFADWSKETRGAPLISVKPLDNWLLIYTRRNYEAANSLIQNLFKVTPAMGMQMRKAIMIEVDDRTEAYLRVLQQKVTADTQIVVCLLSSNRKDKYDAIKKYLCTDCPTPSQCVVARTLGKQQTVMAIATKIALQMNCKMGGELWRVDIPLKLVMIVGIDCYHDMTAGRRSIAGFVASINEGMTRWFSRCIFQDRGQELVDGLKVCLQAALRAWNSCNEYMPSRIIVYRDGVGDGQLKTLVNYEVPQFLDCLKSIGRGYNPRLTVIVVKKRVNTRFFAQSGGRLQNPLPGTVIDVEVTRPEWYDFFIVSQAVRSGSVSPTHYNVIYDNSGLKPDHIQRLTYKLCHIYYNWPGVIRVPAPCQYAHKLAFLVGQSIHREPNLSLSNRLYYL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark E Reeves et al.
PloS one, 9(7), e101679-e101679 (2014-07-10)
RASSF1C is a major isoform of the RASSF1 gene, and is emerging as an oncogene. This is in contradistinction to the RASSF1A isoform, which is an established tumor suppressor. We have previously shown that RASSF1C promotes lung cancer cell proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service