Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1400185

Sigma-Aldrich

Monoclonal Anti-NID1 antibody produced in mouse

clone 1G3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ENTACTIN, Anti-NID, Anti-enactin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NID1(4811)

General description

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix. (provided by RefSeq)

Immunogen

NID1 (NP_002499.1, 1148 a.a. ~ 1247 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EGLQYPFAVTSYGKNLYFTDWKMNSVVALDLAISKETDAFQPHKQTRLYGITTALSQCPQGHNYCSVNNGGCTHLCLATPGSRTCRCPDNTLGVDCIERK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service