Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1400048

Sigma-Aldrich

Anti-CHD2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DKFZp781D1727

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHD2(1106)

General description

The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CHD2 (AAH07347.1, 1 a.a. ~ 501 a.a) full-length human protein.

Sequence
MMRNKDKSQEEDSSLHSNASSHSASEEASGSDSGSQSESEQGSDPGSGHGSESNSSSESSESQSESESESAGSKSQPVLPEAKEKPASKKERIADVKKMWEEYPDVYGVRRSNRSRQEPSRFNIKEEASSGSESGSPKRRGQRQLKKQEKWKQEPSEDEQEQGTSAESEPEQKKVKARRPVPRRTVPKPRVKKQPKTQRGKRKKQDSSDEDDDDDEAPKRQTRRRAAKNVSYKEDDDFETDSDDLIEMTGEGVDEQQDNSETIEKVLDSRLGKKGATGASTTVYAIEANGDPSGDFDTEKDEGEIQYLIKWKGWSYIHSTWESEESLQQQKVKGLKKLENFKKKEDEIKQWLGKVSPEDVEYFNCQQELASELNKQYQIVERVIAVKTSKSTLGQTDFPAHSRKPAPSNEPEYLCKWMGLPYSECSWEDEALIGKKFQNCIDSFHSRNNSKTIPTRECKALKQRPRFVALKKQPAYLGGENLELRDYQLEGLNWLAHSWCK

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service