Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300187

Sigma-Aldrich

Anti-PYGM (C-term) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Glycogen phosphorylase, Anti-Muscle form, Anti-Myophosphorylase, Anti-PYGM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
indirect ELISA: 1:1000
western blot: 1:100-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PYGM(5837)

General description

Glycogen phosphorylase, muscle associated (PYGM) encodes glycogen phosphorylase or myophosphorylase. In human chromosome, the gene PYGM is localized on 11q13.1.

Immunogen

PYGM (NP_005600, 703-737)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human PYGM.

Biochem/physiol Actions

PYGM catalyses and regulates the breakdown of glycogen to glucose-1-phosphate. Defects in PYGM are the cause of Glycogenosis type V/ glycogen storage disease type 5 (GSD5), also known as McArdle disease/ myophosphorylase deficiency, is an autosomal recessive disorder. GSD5 is a metabolic disorder resulting in myopathy characterized by exercise intolerance, cramps, muscle weakness and recurrent myoglobinuria.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of loss of heterozygosity on chromosome 11 and infrequent inactivation of the MEN1 gene in sporadic pituitary adenomas
Tanaka C, et al.
The Journal of Clinical Endocrinology and Metabolism, 83(8), 2631-2634 (1998)
Glycogen storage diseases: diagnosis, treatment and outcome
Chen MA and Weinstein DA
Translational Science of Rare Diseases, 1(1), 45-72 (2016)
Knock-in mice for the R50X mutation in the PYGM gene present with McArdle disease
Nogales-Gadea G, et al.
Brain, 135(7), 2048-2057 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service