Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0110

Sigma-Aldrich

Mouse CTLA-4 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 4.92-1200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ctla4(12477)

General description

The Mouse CTLA4 (cytotoxic T-lymphocyte-associated molecule 4) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse CTLA4 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse CTLA4

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karel Balihar et al.
European journal of gastroenterology & hepatology, 26(8), 880-887 (2014-06-20)
Clostridium difficile infection (CDI) has been increasing in incidence, with significant morbidity and mortality, and is subject to geographical and institutional variability. We aimed to characterize epidemiology and clinical manifestations of CDI in a Czech tertiary care center and to
Amany Mansour et al.
Leukemia & lymphoma, 55(9), 2120-2124 (2013-11-29)
We herein evaluate the role of the B7-family molecule CD86 and the Cytotoxic T-lymphocyte-associated antigen-4 (CTLA-4) as a possible immunopathogenic factors in patients with ALL. The results of 60 patients with de novo ALL were compared to 40 controls. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service