Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0155

Sigma-Aldrich

Rabbit anti-PPP1CC Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

PP-1G, PP1C, PP1gamma, PPP1G, catalytic subunit, gamma isoform, gamma isozyme, protein phosphatase 1, protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_002701.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PPP1CC(5501)

Immunogen

The epitope recognized by PLA0155 maps to a region between residue 280 and the C-terminus (residue 323) of human Protein Phosphatase 1, Catalytic Subunit, Gamma Isoform using the numbering given in entry NP_002701.1 (GeneID 5501).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Protein phosphatase 1 (PP1) is a major eukaryotic serine/threonine phosphatase that is involved in a multitude of cellular functions. PP1 is a holoenzyme that consists of the PP1 catalytic subunit (PP1c) that associates with and is regulated by over 50 regulatory subunits. There are several gene and alternative splice products of the catalytic subunit PP1c. PPP1CC encodes the PP1 gamma catalytic subunit. In addition to PP1 gamma, there are two splice variants of PP1 alpha (PPP1CA), and a PP1 beta (PPP1CB) form. The mutually exclusive interaction between the various catalytic and regulatory subunits allows the PP1 enzyme to be involved in a variety of functions which include cell division, glycogen metabolism, muscle contractility, and protein synthesis. serine/threonine-protein phosphatase PP1-gamma catalytic subunit, PP-1G, PPP1G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanying Hu et al.
PLoS pathogens, 16(7), e1008669-e1008669 (2020-07-24)
Hepatitis B virus (HBV) replicates its genomic DNA via viral DNA polymerase self-primed reverse transcription of a RNA pre-genome in the nucleocapsid assembled by 120 core protein (Cp) dimers. The arginine-rich carboxyl-terminal domain (CTD) of Cp plays an important role
Toshiya Okumura et al.
PloS one, 9(6), e100559-e100559 (2014-06-20)
Phosphorylation of hormone-sensitive lipase (HSL) and perilipin by protein kinase A (PKA) promotes the hydrolysis of lipids in adipocytes. Although activation of lipolysis by PKA has been well studied, inactivation via protein phosphatases is poorly understood. Here, we investigated whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service