Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0013

Sigma-Aldrich

Rabbit anti-ROCK2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ROCK-II, Rho kinase 2, Rho-associated, coiled-coil-containing protein kinase 2, coiled-coil-containing protein kinase II, p164 ROCK-2, rho-associated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:10,000-1:25,000

accession no.

NP_004841.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... ROCK2(9475)

Immunogen

The epitope recognized by PLA0013 maps to a region between residues 1320 and the C-terminus (residues 1388) of human ROCK2 using the number given in entry NP_004841.1 (GeneID 9475)

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Rho-associated protein kinase 2 (ROCK2), or Rho kinase alpha, is a serine-threonine protein kinase that plays an important role in cellular apoptosis, growth, cell motility and cell contraction. ROCK2 shares an overall 65% homology in amino acid sequence with ROCK1. The ROCK1 and ROCK2 isoforms are products of two separate genes. Both ROCK1 and ROCK2 are downstream targets of the Rho A small GTP-binding protein. As downstream effectors of RhoA, ROCK1 and ROCK2 mediate changes in the actin cytoskeleton via phosphorylation of target proteins such as the MBS subunit of MLCP, MLC, ERM proteins, protein LIM kinase and adducin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isidre Ferrer et al.
The Journal of pathology, 233(3), 247-257 (2014-03-08)
Most patients with tuberous sclerosis complex (TSC) develop cortical tubers that cause severe neurological disabilities. It has been suggested that defects in neuronal differentiation and/or migration underlie the appearance of tubers. However, the precise molecular alterations remain largely unknown. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service