Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1534

Sigma-Aldrich

1,2-Didodecanoyl-rac-glycero-3-phosphocholine

≥99%

Synonym(s):

4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxododecyl)oxy]-3,5,9-trioxa-4-phosphaheneicosan-1-aminium 4-oxide, inner salt, rac-1,2-Dilauroyl-glycero-3-phosphocholine, rac-DLPC, rac-Phosphatidylcholine, 1,2-dilauroyl, DL-α-Dilauroyl phosphatidylcholine, DL-β,γ-Dilauroyl-α-lecithin, PC(12:0/12:0)[rac]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H64NO8P
CAS Number:
Molecular Weight:
621.83
Beilstein:
4168394
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCC

InChI

1S/C32H64NO8P/c1-6-8-10-12-14-16-18-20-22-24-31(34)38-28-30(29-40-42(36,37)39-27-26-33(3,4)5)41-32(35)25-23-21-19-17-15-13-11-9-7-2/h30H,6-29H2,1-5H3

InChI key

IJFVSSZAOYLHEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supratik Sen Mojumdar et al.
Langmuir : the ACS journal of surfaces and colloids, 28(27), 10230-10237 (2012-06-19)
Picosecond spectroscopy under a confocal microscope is employed to study solvation dynamics of coumarin 153 (C153) inside a single giant lipid vesicle (1,2-dilauroyl-sn-glycero-3-phosphocholine, DLPC) of diameter 20 μm. Fluorescence correlation spectroscopy (FCS) indicates that the diffusion coefficient (D(t)) of the
Takakuni Oguchi et al.
Journal of oleo science, 60(4), 177-183 (2011-03-24)
The membrane properties of phospholipid mixtures supported on silica were studied by means of a quartz crystal microbalance with dissipation monitoring (QCM-D) technique, in situ soft-contact atomic force microscopy (AFM), and friction force microscopy (FFM). The phospholipids used in this
Michael Weinrich et al.
Langmuir : the ACS journal of surfaces and colloids, 28(10), 4723-4728 (2012-02-23)
X-ray and neutron diffraction studies of a binary lipid membrane demonstrate that halothane at physiological concentrations produces a pronounced redistribution of lipids between domains of different lipid types identified by different lamellar d-spacings and isotope composition. In contrast, dichlorohexafluorocyclobutane (F6)
Matthew E Nipper et al.
Biochimie, 93(6), 988-994 (2011-03-01)
Molecular rotors are a form of fluorescent intramolecular charge-transfer complexes that can undergo intramolecular twisting motion upon photoexcitation. Twisted-state formation leads to non-radiative relaxation that competes with fluorescence emission. In bulk solutions, these molecules exhibit a viscosity-dependent quantum yield. On
Metabolism: A lipid for fat disorders.
Holly A Ingraham
Nature, 474(7352), 455-456 (2011-06-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service