Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6286

Sigma-Aldrich

Anti-Neurogenin 2 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-MATH4A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 28.2 kDa

species reactivity

rodent, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Neurogenin 2 (NEUROG2) is a basic helix loop helix (bHLH) transcription factor.

Immunogen

synthetic peptide corresponding to mouse neurogenin 2 (amino acids 86-98) conjugated to BSA.

Biochem/physiol Actions

Neurogenin 2 (NEUROG2) aids in the differentiation of neuronal tissue in progenitor cells during growth.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penglai Zhao et al.
Brain research, 1615, 51-60 (2015-04-29)
Direct lineage reprogramming of human fibroblasts into functional neurons holds great promise for biomedical applications such as regenerative medicine and cellbased disease modelling. However, clinical applications must consider how to increase neuronal conversion efficiency and at the same time reduce
Sara Gil-Perotin et al.
The European journal of neuroscience, 34(7), 1040-1052 (2011-09-09)
The tumor suppressor protein p53 (Trp53) and the cell cycle inhibitor p27(Kip1) (Cdknb1) have both been implicated in regulating proliferation of adult subventricular zone (aSVZ) cells. We previously reported that genetic ablation of Trp53 (Trp53-/-) or Cdknb1 (p27(Kip1-/-) ) increased
Efil Bayam et al.
BMC genomics, 16, 681-681 (2015-09-06)
Cellular differentiation programs are controlled, to a large extent, by the combinatorial functioning of specific transcription factors. Cortical projection neurons constitute the major excitatory neuron population within the cortex and mediate long distance communication between the cortex and other brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service