Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N3040

Sigma-Aldrich

Necrostatin-7

≥98% (HPLC), powder

Synonym(s):

5-[[3-(4-Fluorophenyl)-1H-pyrazol-4-yl]methylene]-2-imino-3-(2-thiazolyl)-4-thiazolidinone, Nec-7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10FN5OS2
CAS Number:
Molecular Weight:
371.41
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to tan

solubility

DMSO: >10 mg/mL (with warming)

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)-c2n[nH]cc2C=C3SC(=N)N(C3=O)c4nccs4

InChI

1S/C16H10FN5OS2/c17-11-3-1-9(2-4-11)13-10(8-20-21-13)7-12-14(23)22(15(18)25-12)16-19-5-6-24-16/h1-8,18H,(H,20,21)

InChI key

SMJGLNVGTJLIRV-UHFFFAOYSA-N

Biochem/physiol Actions

Necrostatin-7 (Nec-7) is a potent inhibitor of necroptosis, a regulated caspase-independent necrotic cell death pathway, distinct from apoptosis. Nec-7 has a different profile from the other necrostatins as it does not inhibit RIP1 kinase, suggesting the possibility that it may target an additional as yet unknown regulatory molecule in the pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service