Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK087

Sigma-Aldrich

Acid Phosphatase Activity Fluorometric Assay Kit

sufficient for 500 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

usage

sufficient for 500 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

Gene Information

General description

The new Acid Phosphatase Fluorometric Assay Kit, MAK525, is now available! Acid phosphatases (AP) are a family of enzymes that non-specifically catalyze the hydrolysis of monoesters and anhydrides of phosphoric acid to produce inorganic phosphate at an optimal pH of 4-7. Changes in the levels and activities of AP occur in many pathophysiological conditions including prostate cancer and bone disease.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of acid phosphatase in serum and other biological samples.

Principle

The AP Activity Fluorometric Assay Kit provides a simple and direct procedure for measuring AP activity in serum and other samples. AP activity is determined by the conversion of the non-fluorescent 4-methylumbelliferyl phosphate disodium salt (MUP) to the fluorescent product 4-methylumbelliferone (4-MU) (λex = 360/λem = 440 nm), proportional to the AP activity present. One unit of AP is the amount of enzyme that will hydrolyze 1.0 μmole of MUP per minute at 25 °C.

replaced by

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Significance of increased phosphatase activity of bone at the site of osteoplastic metastases secondary to carcinoma of the prostate gland.
Gutman E B, et al.
Cancer Research, 28(3), 485-495 (1936)
Kapil D Patel et al.
ACS applied bio materials, 2(11), 5190-5203 (2019-11-18)
Providing appropriate biophysical and biochemical cues to the interface is a facile strategy to enhance the osteogenic ability of metallic implants. Here we exploited this through the incorporation of mesoporous bioactive glass nanoparticles (MBGN) at a high content (1:1 by
Ruibin Kuang et al.
Plant physiology, 151(1), 199-209 (2009-07-28)
Purple acid phosphatase (PAP) catalyzes the hydrolysis of phosphate monoesters and anhydrides to release phosphate within an acidic pH range. Among the 29 PAP-like proteins in Arabidopsis (Arabidopsis thaliana), AtPAP15 (At3g07130) displays a greater degree of amino acid identity with
Gaurav Datta et al.
Frontiers in cellular neuroscience, 13, 329-329 (2019-08-06)
Human immunodeficiency virus type 1 (HIV-1) associated neuropathy is the most common neurological complication of HIV-1, with debilitating pain affecting the quality of life. HIV-1 gp120 plays an important role in the pathogenesis of HIV neuropathy via direct neurotoxic effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service