Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0289

Sigma-Aldrich

MOPS hemisodium salt

foil pouch

Synonym(s):

3-(N-Morpholino)propanesulfonic acid hemisodium salt, 4-Morpholinepropanesulfonic acid hemisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14.5NO4S · 0.5Na
CAS Number:
Molecular Weight:
220.25
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

form

powder

pH

6.5-7.5 (25 °C)

useful pH range

6.5-7.9

pKa (25 °C)

7.2

SMILES string

[Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN2CCOCC2

InChI

1S/2C7H15NO4S.Na/c2*9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h2*1-7H2,(H,9,10,11);/q;;+1/p-1

InChI key

OVCWZKQPQOVIDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Reconstitution

Contents of each foil pouch dissolved in 1 liter deionized water will yield 0.1 M hemisodium MOPS buffer, pH 7 at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service