Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7151

Sigma-Aldrich

Anti-Insulin-Like Growth Factor-I Receptor antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IGF-I R

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

General description

Insulin-Like Growth Factor-I Receptor (IGF-I receptor) is a disulfide-linked heterotetrameric transmembrane protein that belongs to Tyr protein kinase family and is expressed mainly in all cells and tissues. Association of two α (130 kD) and two β (95 kD) subunits of IGF-I receptor is essential for receptor autophosphorylation. It has a pivotal role in cell growth control as well as can induce differentiation, and eventually cell death, of certain types of cells. Anti-Insulin-Like Growth Factor-I Receptor antibody can be used to block cell surface IGF-I sR mediated IGF-I and IGF-II bioactivity. Goat anti-Insulin-Like Growth Factor-I Receptor antibody reacts specifically with recombinant human IGF-I. The product has shown no cross-reactivity for other cytokines tested.

Specificity

The antibody blocks cell surface IGF-I R mediated IGF-I and IGF-II biological activity. At optimal antibody concentration, no more than 40-60% of the IGF-I or IGF-II activity can be blocked by this antibody.

Immunogen

Recombinant human IGF-I soluble receptor expressed in Sf 21.

Application

Anti-Insulin-Like Growth Factor-I Receptor antibody can be used in indirect ELISA and western blotting.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Baxter et al.
Progress in growth factor research, 1(1), 49-68 (1989-01-01)
Binding proteins for the insulin-like growth factors (IGF-I and IGF-II) are increasingly being recognized as modulators of IGF actions in both inhibitory and stimulatory ways. At least three distinct classes of binding protein are thought to exist, differing in their
The somatomedin-insulin-like growth factors.
K Hall et al.
Journal of internal medicine, 225(1), 47-54 (1989-01-01)
S E Tollefsen et al.
Biochemistry, 30(1), 48-54 (1991-01-08)
We have recently found that association of the two alpha beta dimers of the insulin-like growth factor I (IGF I) receptor is required for formation of a high-affinity binding site for IGF I [Tollefsen, S. E., & Thompson, K. (1988)
R Baserga
Experimental cell research, 253(1), 1-6 (1999-12-02)
The type 1 insulin-like growth factor receptor (IGF-IR) plays an important role in both normal and abnormal growth. It is particularly important in anchorage-independent growth. Impairment of its function causes apoptosis of tumor cells and inhibition of tumor growth in
A M Abbott et al.
The Journal of biological chemistry, 267(15), 10759-10763 (1992-05-25)
The insulin-like growth factor I (IGF I) receptor is a tyrosine kinase-containing transmembrane protein that plays an important role in cell growth control. We have isolated and characterized human genomic DNA clones containing the entire coding sequence of the IGF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service