Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA028465

Sigma-Aldrich

Anti-CRTC2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-CREB regulated transcription coactivator 2, Anti-TORC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SPSASLVLDPPGFSEGPGFLGGEGPMGGPQDPHTFNHQNLTHCSRHGSGPNIILTGDSSPGFSKEIAAALAGVPGFEVSAAGLELGLGL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRTC2(200186)

General description

CREB-regulated transcription co-activator 2 (CRTC2) is also known as TORC (transducer of regulated CREB).

Immunogen

CREB regulated transcription coactivator 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CRTC2 acts as a coactivator for glucocorticoid receptor. CRTC2 stimulates CREB (cAMP response element-binding protein) target gene expression and has a well-established role in modulating glucose and lipid metabolism. It plays an important role in glucagon mediated activation of gluconeogenesis in the early stages of fasting. CRTC2 increases the production of hepatic gluconeogenesis via thyroid stimulating hormone. CRTC2 causes the incidence of metabolic traits in patients after solid organ transplantation. CRCT2 also helps in sterol-responsive element-binding protein 2 (SREPB-2) regulated synthesis of hepatic cholesterol. CRTC2 acts as a lymphoma tumor suppressor gene by activating the transcription of MMR genes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76944

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The CREB Coactivator CRTC2 is a lymphoma tumor suppressor that preserves genome integrity through transcription of DNA mismatch repair genes
Fang, Min, et al.
Testing, 11(9), 1350-1357 (2015)
Clock and light regulation of the CREB coactivator CRTC1 in the suprachiasmatic circadian clock
Sakamoto,, et al.
The Journal of Neuroscience, 33(21), 9021-9027 (2013)
CRTC2 is a coactivator of GR and couples GR and CREB in the regulation of hepatic gluconeogenesis
Hill, Mic, et al.
Molecular Endocrinology, 30(1), 104-117 (2016)
A novel role for CRTC2 in hepatic cholesterol synthesis through SREBP-2
Li, Yujie, et al.
Hepatology, 66(2), 481-497 (2017)
Thyroid stimulating hormone increases hepatic gluconeogenesis via CRTC2
Li, Yujie, et al.
Molecular and Cellular Endocrinology, 446, 70-80 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service