Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA022904

Sigma-Aldrich

Anti-GSTK1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GST 13-13, Anti-GST class-kappa, Anti-GSTK1-1, Anti-Glutathione S-transferase kappa 1, Anti-Glutathione S-transferase subunit 13, Anti-hGSTK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PYSWLGFEILCRYQNIWNINLQLRPSLITGIMKDSGNKPPGLLPRKGLYMANDLKLLRHHLQIPIHFPKDFLSVMLEKGSLSAMRFLTAVNLEHPEMLEKASRELWMRVWSRNE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTK1(373156)

General description

The gene GSTK1 (glutathione S-transferase κ 1) is mapped to human chromosome 7q34. It belongs to the GST superfamily and κ-GST class. The protein is present in the mitochondria and peroxisomes.

Immunogen

Glutathione S-transferase kappa 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Downregulation of GSTK1 impacts cardiac function and contributes to hypertrophic cardiomyopathy. Polymorphisms in GSTK1 gene is implicated in lipid metabolism, obesity and insulin resistance in type 2 diabetes.
GSTs (glutathione S-transferases) are mainly involved in the detoxification pathways and redox biochemistry. GSTK1 (glutathione S-transferase κ 1) shows glutathione conjugating activity towards halogenated aromatics, such as 1-chloro-2,4- dinitrobenzene (CDNB). In addition, it has peroxidase activity with organic hydroperoxides.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70295

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison J Shield et al.
Genomics, 95(5), 299-305 (2010-03-03)
The level of glutathione transferase Kappa (GSTK1-1) has been correlated with obesity (Liu et.al. 2008 PNAS 105: 18302-7) and a polymorphism in the hGSTK1 promoter has been associated with insulin secretion and fat deposition (Gao et al 2009 Endocr J
Dual localization of glutathione S-transferase in the cytosol and mitochondria: implications in oxidative stress, toxicity and disease
Raza H
FEBS Journal, 278(22), 4243-4251 (2011)
Downregulation of GSTK1 is a common mechanism underlying hypertrophic cardiomyopathy
Sasagawa S, et al.
Frontiers in Pharmacology, 7, 162-162 (2016)
Anna Robinson et al.
The Biochemical journal, 379(Pt 3), 541-552 (2004-01-08)
The Kappa class of GSTs (glutathione transferases) comprises soluble enzymes originally isolated from the mitochondrial matrix of rats. We have characterized a Kappa class cDNA from human breast. The cDNA is derived from a single gene comprising eight exons and
Fabrice Morel et al.
The Journal of biological chemistry, 279(16), 16246-16253 (2004-01-27)
Kappa class glutathione S-transferase (GST) cDNA sequences have been identified in rat, mouse, and human. In the present study, we determined the structure and chromosomal location of the human GST Kappa 1 (hGSTK1) gene, characterized the protein, and demonstrated its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service